Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxa13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085494.1 | NP_001078963.1 | 1 - 2 | 786 | 73 | |
GenBank mRNA | BC133882.1 | - | 1 - 2 | 786 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |