Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100063.1 | NP_001093533.1 | 4 - 5 | 589 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |