Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110524.1 | NP_001103994.1 | 4 - 5 | 707 | 96 | |
GenBank mRNA | EU126948.1 | - | 4 - 5 | 707 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Danio rerio Probe spans exons |