Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131801.2 | NP_571876.1 | 3 - 4 | 396 | 69 | |
GenBank mRNA | AB069857.1 | - | 3 - 4 | 396 | 69 | |
AF290981.1 | - | 2 - 3 | 261 | 69 | ||
BC047796.1 | - | 2 - 3 | 361 | 69 | ||
BC063932.1 | - | 3 - 4 | 367 | 69 | ||
BC097034.1 | - | 2 - 3 | 474 | 69 | ||
BC165342.1 | - | 1 - 2 | 133 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |