Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199694.1 | NP_955988.1 | 6 - 7 | 762 | 138 | |
XM_021470452.1 | XP_021326127.1 | 6 - 7 | 824 | 138 | ||
GenBank mRNA | BC062858.1 | - | 6 - 7 | 762 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |