Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lipg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200128.1 | NP_956422.1 | 4 - 5 | 683 | 91 | |
GenBank mRNA | BC044146.1 | - | 4 - 5 | 683 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |