Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200673.1 | NP_956967.1 | 5 - 6 | 518 | 97 | |
XM_005168620.4 | XP_005168677.2 | 5 - 6 | 864 | 97 | ||
GenBank mRNA | BC058063.1 | - | 5 - 6 | 518 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |