Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017621.1 | NP_001017621.1 | 1 - 2 | 202 | 85 | |
GenBank mRNA | BC092828.1 | - | 1 - 2 | 202 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |