Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039805.2 | NP_001034894.2 | 3 - 4 | 404 | 99 | |
GenBank mRNA | AB201385.1 | - | 3 - 4 | 432 | 99 | |
BC083297.1 | - | 3 - 4 | 446 | 99 | ||
BC133981.1 | - | 3 - 4 | 404 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |