Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039982.1 | NP_001035071.1 | 6 - 7 | 1040 | 85 | |
XM_005162709.4 | XP_005162766.1 | 6 - 7 | 953 | 85 | ||
XM_021470220.1 | XP_021325895.1 | 6 - 7 | 1009 | 85 | ||
GenBank mRNA | BC114283.1 | - | 6 - 7 | 1040 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |