Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045162.1 | NP_001038627.1 | 12 - 13 | 4246 | 98 | |
XM_017357360.2 | XP_017212849.1 | 12 - 13 | 4288 | 98 | ||
GenBank mRNA | DQ314737.1 | - | 12 - 13 | 4246 | 98 | |
JQ740885.1 | - | 6 - 7 | 2878 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |