Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045333.2 | NP_001038798.2 | 3 - 4 | 781 | 84 | |
GenBank mRNA | BC118684.1 | - | 3 - 4 | 784 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |