Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clockb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178295.2 | NP_840080.2 | 11 - 12 | 1190 | 99 | |
XM_005168339.4 | XP_005168396.1 | 12 - 13 | 1574 | 99 | ||
GenBank mRNA | AB087256.1 | - | 11 - 12 | 1100 | 99 | |
BC162682.1 | - | 11 - 12 | 1180 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |