Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hes6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194400.2 | NP_919381.2 | 3 - 4 | 320 | 60 | |
GenBank mRNA | AB097830.1 | - | 3 - 4 | 238 | 60 | |
AB158461.1 | - | 3 - 4 | 303 | 60 | ||
BC062387.1 | - | 3 - 4 | 307 | 60 | ||
BC065599.1 | - | 3 - 4 | 320 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |