Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200294.1 | NP_956588.1 | 4 - 5 | 991 | 60 | |
GenBank mRNA | BC049467.1 | - | 4 - 5 | 991 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |