Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chchd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200784.1 | NP_957078.1 | 1 - 2 | 129 | 126 | |
GenBank mRNA | BC059631.1 | - | 1 - 2 | 129 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |