Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007774.1 | NP_001007775.1 | 5 - 6 | 501 | 110 | |
GenBank mRNA | BC085427.1 | - | 5 - 6 | 501 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |