Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031838.1 | NP_001027008.1 | 18 - 19 | 1817 | 82 | |
XM_005161414.4 | XP_005161471.1 | 17 - 18 | 1912 | 82 | ||
XM_005161417.4 | XP_005161474.1 | 18 - 19 | 1807 | 82 | ||
GenBank mRNA | AY918066.1 | - | 18 - 19 | 1817 | 82 | |
BC057511.1 | - | 17 - 18 | 1798 | 82 | ||
BC133108.1 | - | 18 - 19 | 1721 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |