Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076772.1 | NP_001070240.1 | 1 - 2 | 750 | 59 | |
GenBank mRNA | BC124165.1 | - | 1 - 2 | 750 | 59 | |
BC165512.1 | - | 1 - 2 | 488 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |