Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 6 - 7 | 808 | 79 | |
XM_005168796.4 | XP_005168853.1 | 6 - 7 | 3509 | 79 | ||
GenBank mRNA | BC066564.1 | - | 6 - 7 | 808 | 79 | |
BC129198.1 | - | 6 - 7 | 808 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |