Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pyya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164371.1 | NP_001157843.1 | 2 - 3 | 331 | 67 | |
NM_131016.2 | NP_571091.1 | 1 - 2 | 267 | 67 | ||
GenBank mRNA | BC162428.1 | - | 1 - 2 | 267 | 67 | |
BC162436.1 | - | 1 - 2 | 267 | 67 | ||
EH440265.1 | - | 2 - 3 | 362 | 67 | ||
FJ915073.1 | - | 1 - 2 | 178 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |