Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002357.1 | NP_001002357.1 | 3 - 4 | 344 | 112 | |
XM_021466675.1 | XP_021322350.1 | 1 - 2 | 951 | 112 | ||
GenBank mRNA | BC075898.1 | - | 3 - 4 | 344 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |