Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013274.1 | NP_001013292.1 | 2 - 3 | 360 | 92 | |
XM_005164742.3 | XP_005164799.1 | 2 - 3 | 1286 | 92 | ||
GenBank mRNA | BC135005.1 | - | 2 - 3 | 363 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |