Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024436.1 | NP_001019607.1 | 5 - 6 | 906 | 75 | |
GenBank mRNA | BC076161.1 | - | 5 - 6 | 793 | 75 | |
BC095058.1 | - | 5 - 6 | 906 | 75 | ||
KT008427.1 | - | 5 - 6 | 747 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |