Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025523.1 | NP_001020694.1 | 3 - 4 | 454 | 84 | |
XM_005160740.4 | XP_005160797.1 | 3 - 4 | 955 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |