Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025551.1 | NP_001020722.1 | 8 - 9 | 2158 | 97 | |
XM_005160586.3 | XP_005160643.1 | 9 - 10 | 2727 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |