Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mhc1uba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131471.1 | NP_571546.1 | 2 - 3 | 386 | 61 | |
GenBank mRNA | BC074095.1 | - | 2 - 3 | 360 | 61 | |
BC164159.1 | - | 2 - 3 | 347 | 61 | ||
Z46777.1 | - | 386 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |