Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mhc1uba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131471.1 | NP_571546.1 | 5 - 6 | 1050 | 127 | |
GenBank mRNA | BC074095.1 | - | 5 - 6 | 1024 | 127 | |
Z46777.1 | - | 1050 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |