Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199832.2 | NP_956126.1 | 5 - 6 | 484 | 60 | |
GenBank mRNA | AY192987.1 | - | 5 - 6 | 397 | 60 | |
BC044148.1 | - | 5 - 6 | 469 | 60 | ||
BC164324.1 | - | 5 - 6 | 431 | 60 | ||
EH580836.1 | - | 5 - 6 | 495 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |