Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igfbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205751.2 | NP_991314.2 | 3 - 4 | 947 | 71 | |
GenBank mRNA | AJ299410.1 | - | 3 - 4 | 942 | 71 | |
BC081574.1 | - | 3 - 4 | 947 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |