Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcnj11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012387.2 | NP_001012387.1 | 4 - 5 | 820 | 62 |
Gene Symbol: LOC103909814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC108004.1 | - | 4 - 5 | 820 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |