Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03147629_m1
See other DLG1L GE Assays ›
Gene Symbol:
dlg1l
Gene Name:
discs, large (Drosophila) homolog 1, like
Gene Aliases:
-
Chromosome Location:
Chr.2: 4902770 - 5074670 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03147629_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: dlg1l
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001012388.1
NP_001012388.1
6 - 7
653
59
XM_005163667.4
XP_005163724.1
9 - 10
1554
59
XM_005163668.4
XP_005163725.1
9 - 10
1536
59
XM_005163669.4
XP_005163726.1
9 - 10
1533
59
XM_005163671.4
XP_005163728.1
8 - 9
1463
59
XM_005163673.4
XP_005163730.1
8 - 9
1437
59
XM_005163675.4
XP_005163732.1
8 - 9
1463
59
XM_005163676.4
XP_005163733.1
7 - 8
1364
59
XM_005163677.4
XP_005163734.1
7 - 8
1363
59
XM_005163678.4
XP_005163735.1
5 - 6
1181
59
XM_005163682.4
XP_005163739.1
4 - 5
468
59
XM_009296932.3
XP_009295207.1
10 - 11
1654
59
XM_009296934.3
XP_009295209.1
10 - 11
1653
59
XM_009296940.3
XP_009295215.1
9 - 10
1581
59
XM_009296951.3
XP_009295226.1
10 - 11
1653
59
XM_009296964.3
XP_009295239.1
7 - 8
1369
59
XM_009296971.3
XP_009295246.1
6 - 7
1249
59
XM_009296979.3
XP_009295254.1
4 - 5
606
59
XM_009296993.3
XP_009295268.1
5 - 6
1178
59
XM_021480105.1
XP_021335780.1
10 - 11
1651
59
XM_021480123.1
XP_021335798.1
7 - 8
806
59
XM_021480126.1
XP_021335801.1
4 - 5
1223
59
GenBank mRNA
AY819035.1
-
6 - 7
653
59
BC163455.1
-
6 - 7
631
59
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Mammalian Gene Collection
Recommended Assay
Danio rerio
Probe spans exons
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
endothelial cell proliferation
actin filament organization
chemical synaptic transmission
embryo development
cortical actin cytoskeleton organization
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
negative regulation of mitotic cell cycle
receptor localization to synapse
cell-cell adhesion
neurotransmitter receptor localization to postsynaptic specialization membrane
protein kinase binding
phosphatase binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0