Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc6a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039972.1 | NP_001035061.1 | 10 - 11 | 1677 | 71 | |
XM_005157654.4 | XP_005157711.1 | 13 - 14 | 2855 | 71 | ||
XM_005157655.4 | XP_005157712.1 | 13 - 14 | 2778 | 71 | ||
XM_009291668.3 | XP_009289943.1 | 14 - 15 | 3047 | 71 | ||
XM_009291669.3 | XP_009289944.1 | 14 - 15 | 2965 | 71 | ||
XM_009291670.3 | XP_009289945.1 | 12 - 13 | 2586 | 71 | ||
XM_009291671.3 | XP_009289946.1 | 14 - 15 | 2343 | 71 | ||
XM_009291672.3 | XP_009289947.1 | 12 - 13 | 2504 | 71 | ||
XM_009291673.3 | XP_009289948.1 | 10 - 11 | 1426 | 71 | ||
GenBank mRNA | BC163766.1 | - | 10 - 11 | 1677 | 71 | |
BC163777.1 | - | 10 - 11 | 1677 | 71 | ||
DQ285098.1 | - | 10 - 11 | 1677 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |