Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amotl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080177.1 | NP_001073646.1 | 9 - 10 | 2132 | 65 | |
GenBank mRNA | BC163299.1 | - | 9 - 10 | 2006 | 65 | |
DQ887096.1 | - | 9 - 10 | 2132 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |