Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131565.1 | NP_571640.1 | 2 - 3 | 66 | 62 | |
XM_005163151.3 | XP_005163208.3 | 3 - 4 | 206 | 62 | ||
GenBank mRNA | AF180889.1 | - | 2 - 3 | 66 | 62 | |
BC053304.1 | - | 3 - 4 | 89 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |