Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200226.2 | NP_956520.2 | 7 - 8 | 858 | 88 | |
GenBank mRNA | AY391457.1 | - | 6 - 7 | 657 | 88 | |
BC046061.1 | - | 7 - 8 | 684 | 88 | ||
BC071375.1 | - | 7 - 8 | 858 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |