Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxl14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201482.1 | NP_958890.1 | 1 | 952 | 90 | |
GenBank mRNA | AF467461.1 | - | 1 | 559 | 90 | |
AY398381.1 | - | 1 | 952 | 90 | ||
BC096775.1 | - | 1 | 887 | 90 | ||
BC153539.1 | - | 2 | 679 | 90 | ||
BC164514.1 | - | 1 | 608 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |