Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213154.1 | NP_998319.1 | 5 - 6 | 320 | 135 | |
GenBank mRNA | BC053233.1 | - | 5 - 6 | 320 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |