Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002531.1 | NP_001002531.1 | 5 - 6 | 546 | 111 | |
GenBank mRNA | BC076365.1 | - | 5 - 6 | 546 | 111 | |
BC165890.1 | - | 3 - 4 | 368 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |