Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003856.1 | NP_001003856.1 | 1 | 355 | 85 | |
GenBank mRNA | AY226101.1 | - | 1 | 355 | 85 | |
BC162102.1 | - | 1 | 354 | 85 | ||
BC162473.1 | - | 1 | 354 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |