Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003890.3 | NP_001003890.3 | 4 - 5 | 1061 | 141 | |
GenBank mRNA | AY648752.1 | - | 4 - 5 | 141 | ||
BC098545.1 | - | 4 - 5 | 1061 | 141 | ||
HM210100.1 | - | 4 - 5 | 777 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |