Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004577.1 | NP_001004577.1 | 7 - 8 | 1450 | 61 | |
GenBank mRNA | BC081650.1 | - | 7 - 8 | 1450 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |