Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004680.1 | NP_001004680.1 | 3 - 4 | 300 | 80 | |
GenBank mRNA | BC081493.1 | - | 3 - 4 | 300 | 80 | |
BC164442.1 | - | 2 - 3 | 250 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |