Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005949.1 | NP_001005949.1 | 5 - 6 | 719 | 86 | |
GenBank mRNA | BC083455.1 | - | 5 - 6 | 719 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |