Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013573.1 | NP_001013591.1 | 8 - 9 | 1018 | 60 | |
XM_021481013.1 | XP_021336688.1 | 8 - 9 | 1045 | 60 | ||
XM_021481015.1 | XP_021336690.1 | 8 - 9 | 1045 | 60 | ||
XM_021481016.1 | XP_021336691.1 | 8 - 9 | 1067 | 60 | ||
GenBank mRNA | BC090463.1 | - | 8 - 9 | 1018 | 60 | |
BC154362.1 | - | 8 - 9 | 1039 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |