Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017879.1 | NP_001017879.1 | 4 - 5 | 503 | 58 | |
GenBank mRNA | BC092969.1 | - | 4 - 5 | 503 | 58 | |
BC158193.1 | - | 4 - 5 | 516 | 58 | ||
BC165503.1 | - | 4 - 5 | 390 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |