Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnaset2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030064.2 | NP_001025235.2 | 5 - 6 | 428 | 109 | |
NM_001320391.1 | NP_001307320.1 | 4 - 5 | 407 | 109 | ||
XM_021480672.1 | XP_021336347.1 | 5 - 6 | 447 | 109 | ||
GenBank mRNA | BC096907.1 | - | 5 - 6 | 397 | 109 | |
BC165927.1 | - | 3 - 4 | 213 | 109 | ||
FJ460212.1 | - | 4 - 5 | 376 | 109 | ||
KM655669.1 | - | 3 - 4 | 179 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |