Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002041.1 | NP_001002041.1 | 2 - 3 | 249 | 156 | |
GenBank mRNA | AY648765.1 | - | 2 - 3 | 249 | 156 | |
BC045326.1 | - | 2 - 3 | 250 | 156 | ||
BC163908.1 | - | 2 - 3 | 238 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |