Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002181.1 | NP_001002181.1 | 2 - 3 | 429 | 113 | |
GenBank mRNA | BC074043.1 | - | 2 - 3 | 429 | 113 | |
BC155262.1 | - | 2 - 3 | 368 | 113 | ||
DQ100356.1 | - | 2 - 3 | 364 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |