Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044343.2 | NP_001037808.2 | 18 - 19 | 2145 | 65 | |
XM_005162427.4 | XP_005162484.2 | 18 - 19 | 2089 | 65 | ||
GenBank mRNA | AI544602.1 | - | 1 - 2 | 398 | 65 | |
BC133839.1 | - | 18 - 19 | 2122 | 65 | ||
BC163451.1 | - | 18 - 19 | 65 | |||
DQ019227.1 | - | 18 - 19 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |